accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3ClO

CAS RN 80443-63-6

IUPAC Name: 2-tert-butyl-2-[2-(4-chlorophenyl)ethyl]oxirane
Synonyms: 2-tert-butyl-2-[2-(4-chlorophenyl)ethyl]oxirane
Molecular Weight (g/mol): 238.76
Molecular Formula: C14H19ClO
InChi Key: QLCGXXYDHCTVKP-UHFFFAOYNA-N
SMILES: CC(C)(C)C1(CCC2=CC=C(Cl)C=C2)CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results