accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80500-27-2

IUPAC Name: (4-methyl-3-nitrophenyl)boronic acid
Synonyms: 4-methyl-3-nitrophenylboronic acid
Molecular Weight (g/mol): 180.95
Molecular Formula: C7H8BNO4
InChi Key: OASVXBRTNVFKFS-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1[N+]([O-])=O)B(O)O
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results