Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-3-nitrophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M11275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1127-5G 5 g
M11271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1127-5G Supplier TCI America Supplier No. M11275G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 80500-27-2
Molecular Formula C7H8BNO4
Quantity 5 g
MDL Number MFCD00191550
Synonym 4-methyl-3-nitrophenyl boronic acid, 4-methyl-3-nitrobenzeneboronic acid, 3-nitro-p-tolylboronic acid, 4-methyl-3-nitrophenyl boranediol, 4-methyl-3-nitro-phenyl boronic acid
InChI Key OASVXBRTNVFKFS-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)C)[N+](=O)[O-])(O)O
IUPAC Name (4-methyl-3-nitrophenyl)boronic acid
Molecular Weight (g/mol) 180.954
PubChem CID 2773515
Formula Weight 180.95
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.