accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3CH3H3COHSOOO

CAS RN 80526-82-5

IUPAC Name: tetramethylazanium hydrogen sulfate
Synonyms: tetramethylammonium ion hydrogen sulfate
Molecular Weight (g/mol): 171.21
Molecular Formula: C4H13NO4S
InChi Key: DWTYPCUOWWOADE-UHFFFAOYSA-M
SMILES: C[N+](C)(C)C.OS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)