Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium Hydrogen Sulfate 96.0+%, TCI America™
SDP

Catalog No. T084025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T084025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T084025G Supplier TCI America Supplier No. T084025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 80526-82-5
Molecular Formula C4H13NO4S
Molecular Weight (g/mol) 171.211
MDL Number MFCD00036149
InChI Key DWTYPCUOWWOADE-UHFFFAOYSA-M
Synonym tetramethylammonium hydrogen sulfate, tetramethylammonium bisulfate, tetramethylammonium hydrogen sulphate, tetramethylammonium hydrogensulfate, hydrogen sulfate; tetramethylazanium, methanaminium, n,n,n-trimethyl-, sulfate 1:1, hydrogen sulfate; tetramethylammonium, acmc-209pjn, ch3 4n hso4 .h2o, tetramethylammonium bisulfate solution
PubChem CID 157340
IUPAC Name hydrogen sulfate;tetramethylazanium
SMILES C[N+](C)(C)C.OS(=O)(=O)[O-]

Specifications

Color White
Quantity 25 g
Formula Weight 171.21
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylammonium Hydrogen Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.