CAS RN 8061-51-6
IUPAC Name:
disodium 3-(2-hydroxy-3-methoxyphenyl)-2-[2-methoxy-4-(3-sulfonatopropyl)phenoxy]propane-1-sulfonate
Synonyms:
sodium lignosulfonate
Molecular Weight (g/mol):
534.5
Molecular Formula:
C20H24Na2O10S2
InChi Key:
YDEXUEFDPVHGHE-UHFFFAOYNA-L
SMILES:
[Na+].[Na+].COC1=CC=CC(CC(CS([O-])(=O)=O)OC2=C(OC)C=C(CCCS([O-])(=O)=O)C=C2)=C1O
1
–
1
of
1
results
1
Sigma Aldrich Lignosulfonic acid sodium salt
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.