accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80826-86-4

IUPAC Name: 4-iodo-3,5-dimethylphenol
Synonyms: 4-iodo-3,5-dimethylphenol
Molecular Weight (g/mol): 248.06
Molecular Formula: C8H9IO
InChi Key: RMJZLNUEDDXELD-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC(C)=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results