Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodo-3,5-dimethylphenol, 98%, Thermo Scientific™

Catalog No. AAA1729203
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1729203 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1729203 Supplier Thermo Scientific Chemicals Supplier No. A1729203

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 80826-86-4
Molecular Formula C8H9IO
Quantity 1g
MDL Number MFCD00174284
Sensitivity Light sensitive
Synonym 3,5-dimethyl-4-iodophenol, 4-iodo-3,5-dimethyl-phenol, pubchem19272, 3,5-xylenol,4-iodo, 5-hydroxy-2-iodo-m-xylene, phenol,4-iodo-3,5-dimethyl, 3,5-dimethyl-4-iodo phenol
InChI Key RMJZLNUEDDXELD-UHFFFAOYSA-N
SMILES CC1=CC(=CC(=C1I)C)O
IUPAC Name 4-iodo-3,5-dimethylphenol
Molecular Weight (g/mol) 248.063
PubChem CID 2736264
Formula Weight 248.06
Percent Purity 98%
Chemical Name or Material 4-Iodo-3,5-dimethylphenol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.