accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3HOCl

CAS RN 80854-14-4

IUPAC Name: 2-(4-chlorophenyl)-2-methylpropan-1-ol
Synonyms: 2-(4-chlorophenyl)-2-methylpropan-1-ol
Molecular Weight (g/mol): 184.66
Molecular Formula: C10H13ClO
InChi Key: NJTICDHSNUXDFF-UHFFFAOYSA-N
SMILES: CC(C)(CO)C1=CC=C(Cl)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results