Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Chlorophenyl)-2-methylpropanol 98.0+%, TCI America™
SDP

Catalog No. C21765G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C21765G Supplier TCI America Supplier No. C21765G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 80854-14-4
Molecular Formula C10H13ClO
Molecular Weight (g/mol) 184.66
MDL Number MFCD06657569
InChI Key NJTICDHSNUXDFF-UHFFFAOYSA-N
PubChem CID 13034557
IUPAC Name 2-(4-chlorophenyl)-2-methylpropan-1-ol
SMILES CC(C)(CO)C1=CC=C(Cl)C=C1

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 5 g
Formula Weight 184.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Chlorophenyl)-2-methylpropanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.