accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80866-74-6

IUPAC Name: hydrogen 2-azatricyclo[10.4.0.0⁴,⁹]hexadeca-1(16),4,6,8,12,14-hexaene chloride
Synonyms: hydrogen 2-azatricyclo[10.4.0.0⁴,⁹]hexadeca-1(16),4,6,8,12,14-hexaene chloride
Molecular Weight (g/mol): 245.75
Molecular Formula: C15H16ClN
InChi Key: UOKRHEQKLWSMMM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C1CC2=CC=CC=C2NCC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results