Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6,11,12-Tetrahydrodibenz[b,f]azocine Hydrochloride, Thermo Scientific™

Catalog No. AC182080010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC182080010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC182080010 Supplier Thermo Scientific Chemicals Supplier No. 182080010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80866-74-6
Molecular Formula C15H16ClN
Molecular Weight (g/mol) 245.75
InChI Key UOKRHEQKLWSMMM-UHFFFAOYSA-N
Synonym 5,6,11,12-tetrahydrodibenz b,f azocinium chloride, 5,6,11,12-tetrahydrodibenz b,f azocine hydrochloride, 5,6,11,-12-tetrahydrodibenz b,f azocine hydrochloride, 5,6,11,12-tetrahydrobenzo c 1 benzazocine hydrochloride, 2-azatricyclo 10.4.0.0?,? hexadeca-1 12 ,4 9 ,5,7,13,15-hexaene hydrochloride, 2-azatricyclo 10.4.0.0?,? hexadeca-1 16 ,4,6,8,12,14-hexaene hydrochloride
PubChem CID 2724014
IUPAC Name 5,6,11,12-tetrahydrobenzo[c][1]benzazocine;hydrochloride
SMILES C1CC2=CC=CC=C2NCC3=CC=CC=C31.Cl

Specifications

Quantity 1 g
Formula Weight 245.76
Chemical Name or Material 5,6,11,12-Tetrahydrodibenz[b,f]azocine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.