accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNOO

CAS RN 80866-76-8

IUPAC Name: (3-methyl-2-nitrophenyl)methanol
Synonyms: (3-methyl-2-nitrophenyl)methanol
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: NELPTBUSFQMYOB-UHFFFAOYSA-N
SMILES: CC1=CC=CC(CO)=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results