accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH2NONH2O

CAS RN 80866-90-6

IUPAC Name: 2-ethyl-2-phenylpropanediamide
Synonyms: 2 ethyl 2 phenylmalonamide
Molecular Weight (g/mol): 206.25
Molecular Formula: C11H14N2O2
InChi Key: JFZHPFOXAAIUMB-UHFFFAOYSA-N
SMILES: CCC(C(N)=O)(C(N)=O)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results