Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-2-phenylmalonamide monohydrate, 99%, Thermo Scientific™

Catalog No. AC187112500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC187112500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC187112500 Supplier Thermo Scientific Chemicals Supplier No. 187112500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80866-90-6
Molecular Formula C11H16N2O3
Molecular Weight (g/mol) 224.26
InChI Key FZSBWHZDTUPYRX-UHFFFAOYSA-N
Synonym 2-ethyl-2-phenylmalonamide monohydrate, unii-27l1min7yr, phenylethylmalonamide hydrate, 27l1min7yr, propanediamide, 2-ethyl-2-phenyl-, hydrate 1:1, 2-ethyl-2-phenylmalonamide hydrate, 2 ethyl 2 phenylmalonamide hydrate, ethylphenylmalonamide hydrate, propanediamide, 2-ethyl-2-phenyl-, monohydrate, acmc-1bl78
PubChem CID 2724041
IUPAC Name 2-ethyl-2-phenylpropanediamide;hydrate
SMILES CCC(C1=CC=CC=C1)(C(=O)N)C(=O)N.O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula C6H5C(C2H5)(CONH2)2·H2O
Quantity 250 mg
Formula Weight 224.26
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Ethyl-2-phenylmalonamide monohydrate, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.