accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOCH3

CAS RN 80913-66-2

IUPAC Name: 4-methylmorpholin-4-ium-4-olate
Synonyms: methyl morpholine oxide
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: LFTLOKWAGJYHHR-UHFFFAOYSA-N
SMILES: C[N+]1([O-])CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results