accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80970-09-8

HClH3CCH3NH2NH2O

CAS RN 80970-09-8

IUPAC Name: 2-amino-4-methylpentanamide hydrochloride
Synonyms: 2-amino-4-methylpentanamide hydrochloride
Molecular Weight (g/mol): 166.65
Molecular Formula: C6H15ClN2O
InChi Key: VSPSRRBIXFUMOU-UHFFFAOYNA-N
SMILES: Cl.CC(C)CC(N)C(N)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)