accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3NOOH3CCH3H3CNOONOO

CAS RN 81-15-2

IUPAC Name: 1-tert-butyl-3,5-dimethyl-2,4,6-trinitrobenzene
Synonyms: musk xylene
Molecular Weight (g/mol): 297.27
Molecular Formula: C12H15N3O6
InChi Key: XMWRWTSZNLOZFN-UHFFFAOYSA-N
SMILES: CC1=C(C(=C(C(C)=C1[N+]([O-])=O)[N+]([O-])=O)C(C)(C)C)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results