Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trinitro-1-tert-butyl-3,5-dimethylbenzene, MP Biomedicals

Catalog No. ICN204822 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204822 Supplier MP Biomedicals, Inc Supplier No. 0520482280

Chemical Identifiers

CAS 81-15-2
Molecular Formula C12H15N3O6
Molecular Weight (g/mol) 297.267
InChI Key XMWRWTSZNLOZFN-UHFFFAOYSA-N
Synonym musk xylene, musk xylol, xylene musk, 2,4,6-trinitro-1,3-dimethyl-5-tert-butylbenzene, 5-tert-butyl-2,4,6-trinitro-m-xylene, 5-tert-butyl-2,4,6-trinitroxylene, 2,4,6-trinitro-5-tert-butyl-m-xylene, m-xylene, 5-tert-butyl-2,4,6-trinitro, unii-1zao16gu5k, ccris 2391
PubChem CID 62329
ChEBI CHEBI:77320
IUPAC Name 1-tert-butyl-3,5-dimethyl-2,4,6-trinitrobenzene
SMILES CC1=C(C(=C(C(=C1[N+](=O)[O-])C(C)(C)C)[N+](=O)[O-])C)[N+](=O)[O-]

Specifications

Quantity 100 g
Chemical Name or Material 2,4,6-Trinitro-1-Tert-Butyl-3,5-Dimethylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.