accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClCl

CAS RN 81-19-6

IUPAC Name: 1,3-dichloro-2-(dichloromethyl)benzene
Synonyms: 1,3-dichloro-2-(dichloromethyl)benzene
Molecular Weight (g/mol): 229.91
Molecular Formula: C7H4Cl4
InChi Key: QQPXXHAEIGVZKQ-UHFFFAOYSA-N
SMILES: ClC(Cl)C1=C(Cl)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results