Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha,2,6-Tetrachlorotoluene 98.0+%, TCI America™
SDP

Catalog No. T007325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T007325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T007325G Supplier TCI America Supplier No. T007325G

Chemical Identifiers

CAS 81-19-6
Molecular Formula C7H4Cl4
Molecular Weight (g/mol) 229.909
MDL Number MFCD00016349
InChI Key QQPXXHAEIGVZKQ-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzal chloride, alpha,alpha,2,6-tetrachlorotoluene, 1,3-dichloro-2-dichloromethyl benzene, 2,6-dichlorobenzylidene chloride, benzene, 1,3-dichloro-2-dichloromethyl, alpha,alpha2,6-tetrachlorotoluene, alpha,alpha-2,6-tetrachlorotoluene, unii-7p4lg60h4s, toluene, .alpha.,.alpha.2,6-tetrachloro, acmc-1bkgm
PubChem CID 6671
IUPAC Name 1,3-dichloro-2-(dichloromethyl)benzene
SMILES C1=CC(=C(C(=C1)Cl)C(Cl)Cl)Cl

Specifications

Color Yellow
Boiling Point 250°C
Quantity 25 g
UN Number 1760
Formula Weight 229.91
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha,alpha,2,6-Tetrachlorotoluene
EINECSNumber (3)-0103
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.