CAS RN 81-25-4
IUPAC Name:
(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Synonyms:
cholic acid 胆酸 cholsaeure
Molecular Weight (g/mol):
408.58
Molecular Formula:
C24H40O5
InChi Key:
BHQCQFFYRZLCQQ-OELDTZBJSA-N
SMILES:
C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
1
–
2
of
2
results
1
Promotions Available
Cholic acid, 97%, Thermo Scientific Chemicals
Cholic acid, 97%, Thermo Scientific Chemicals
CAS: 81-25-4 Molecular Formula: C24H40O5 Molecular Weight (g/mol): 408.579 MDL Number: MFCD00003672 InChI Key: BHQCQFFYRZLCQQ-OELDTZBJSA-N Synonym: cholic acid,cholalic acid,cholate,cholalin,colalin,cholsaeure,cholbam,cholicacid,cholic acid, 5beta,3alpha,7alpha,12alpha-trihydroxy-5beta-cholanic acid PubChem CID: 221493 ChEBI: CHEBI:16359 IUPAC Name: (4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid SMILES: CC(CCC(=O)O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C