Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholic acid, MP Biomedicals™

Catalog No. ICN10138391 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10138391 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10138391 Supplier MP Biomedicals, Inc Supplier No. 0210138391
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 81-25-4
Color White
Assay Percent Range 99.6%
Molecular Formula C24H40O5
Quantity 1 kg
MDL Number MFCD00003672
Synonym cholic acid, cholalic acid, cholate, cholalin, colalin, cholsaeure, cholbam, cholicacid, cholic acid, 5beta, 3alpha,7alpha,12alpha-trihydroxy-5beta-cholanic acid
InChI Key BHQCQFFYRZLCQQ-OELDTZBJSA-N
SMILES CC(CCC(=O)O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
IUPAC Name (4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molecular Weight (g/mol) 408.579
PubChem CID 221493
ChEBI CHEBI:16359
Formula Weight 408.58
Physical Form Powder
Chemical Name or Material Cholic acid, from Bovine or Ovine Bile
Show More Show Less
EINECSNumber 201-337-8
Recommended Storage Store at Room Temperature (15 to 30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.