accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOOO

CAS RN 81-81-2

IUPAC Name: 2-hydroxy-3-(3-oxo-1-phenylbutyl)-4H-chromen-4-one
Synonyms: coumarins warfarin, sodium deriv. warficide athrombin-k athrombin-K maag rattentod cum caswell no. 903 rodafarin eastern states duocide D-con
Molecular Weight (g/mol): 308.33
Molecular Formula: C19H16O4
InChi Key: QTXVAVXCBMYBJW-UHFFFAOYNA-N
SMILES: CC(=O)CC(C1=CC=CC=C1)C1=C(O)OC2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results