Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-3-(α-Acetonylbenzyl)-4-hydroxycoumarin, MP Biomedicals

Catalog No. ICN211609 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211609 Supplier MP Biomedicals, Inc Supplier No. 0521160910

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 81-81-2
Molecular Formula C19H16O4
Molecular Weight (g/mol) 308.333
InChI Key PJVWKTKQMONHTI-UHFFFAOYSA-N
Synonym warfarin, coumadin, coumafene, prothromadin, coumafen, coumarins, zoocoumarin, coumefene, panwarfin, warfarine
PubChem CID 54678486
ChEBI CHEBI:87732
IUPAC Name 4-hydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one
SMILES CC(=O)CC(C1=CC=CC=C1)C2=C(C3=CC=CC=C3OC2=O)O

Specifications

Quantity 10 g
Formula Weight 308.32
Grade Pure
Chemical Name or Material DL-3-(α-Acetonylbenzyl)-4-hydroxycoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.