accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHOHOO

CAS RN 81-90-3

IUPAC Name: 2-[bis(4-hydroxyphenyl)methyl]benzoic acid
Synonyms: phenolphthalin
Molecular Weight (g/mol): 320.34
Molecular Formula: C20H16O4
InChi Key: FFFPYJTVNSSLBQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
Color 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results