accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3H3CCH3HOOSOOH3CCH3OCH3

CAS RN 81012-93-3

IUPAC Name: sodium 4-{3-[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl}-5-methyl-2-(propan-2-yl)benzen-1-olate
Synonyms: sodium 4-[3-(4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxo-2,1λ⁶-benzoxathiol-3-yl]-2-isopropyl-5-methylbenzenolate
Molecular Weight (g/mol): 488.57
Molecular Formula: C27H29NaO5S
InChi Key: BAVBEHWEOJMHDS-UHFFFAOYNA-M
SMILES: [Na+].CC(C)C1=CC(=C(C)C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C([O-])C=C1C
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)