accessibility menu, dialog, popup

UserName

Viewing profile as user:
OClOOOOClOOOOClOOOGa

CAS RN 81029-07-4

IUPAC Name: gallium(3+) triperchlorate
Synonyms: gallium(3+) triperchlorate
Molecular Weight (g/mol): 368.06
Molecular Formula: Cl3GaO12
InChi Key: DKRHELWBVMBPOQ-UHFFFAOYSA-K
SMILES: [Ga+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)