Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium(III) perchlorate hydrate, 99%, Thermo Scientific™

Catalog No. AC196920010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC196920010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC196920010 Supplier Thermo Scientific Chemicals Supplier No. 196920010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81029-07-4
Molecular Formula Cl3GaO12
Molecular Weight (g/mol) 368.06
MDL Number MFCD00016099
InChI Key DKRHELWBVMBPOQ-UHFFFAOYSA-K
Synonym gallium iii perchlorate hydrate, gallium perchlorate-water 1/3/1, gallium 3+ hydrate triperchlorate, gallium perchlorate hydrate, gallium iii perchlorate hydrate ga, gallium iii perchlorate hydrate trace metals basis
PubChem CID 16217276
IUPAC Name gallium(3+) triperchlorate
SMILES [Ga+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Color White
Physical Form Adhering Crystals
Quantity 1 g
Assay Percent Range 99%
Linear Formula Ga(ClO4)3
Formula Weight 368.07
Percent Purity ≥98.5%
Chemical Name or Material Gallium(III) perchlorate hydrate, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.