CAS RN 810659-53-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Bcl-2 Inhibitor II, YC137
The Bcl-2 Inhibitor II, YC137, also referenced under CAS 810659-53-1, controls the biological activity of Bcl-2. This small molecule/inhibitor is primarily used for Activators/Inducers applications.