Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bcl-2 Inhibitor II, YC137

Catalog No. 1973315MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-733-15MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-733-15MG Supplier MilliporeSigma™ Supplier No. 1973315MG
Add to Cart
Add to Cart

The Bcl-2 Inhibitor II, YC137, also referenced under CAS 810659-53-1, controls the biological activity of Bcl-2. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Description 90% by HPLC
Target Proteins Bcl-2
CAS 810659-53-1
Molecular Formula C24H21N3O6S2
Formula Weight 511.6g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.