accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 81156-68-5

IUPAC Name: 2-(2,4-dichlorophenyl)ethan-1-ol
Synonyms: 2-(2,4-dichlorophenyl)ethanol
Molecular Weight (g/mol): 191.05
Molecular Formula: C8H8Cl2O
InChi Key: JIJGKPVJAALUQQ-UHFFFAOYSA-N
SMILES: OCCC1=CC=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results