Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,4-Dichlorophenyl)ethanol 97.0+%, TCI America™
SDP

Catalog No. D29215G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29215G Supplier TCI America Supplier No. D29215G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 81156-68-5
Molecular Formula C8H8Cl2O
Molecular Weight (g/mol) 191.051
MDL Number MFCD00192010
InChI Key JIJGKPVJAALUQQ-UHFFFAOYSA-N
Synonym 2,4-dichlorophenethyl alcohol, 2-2,4-dichlorophenyl ethanol, 2,4-dichlorophenethylalcohol, 2-2,4-dichlorophenyl ethan-1-ol, 2,4-dichlorobenzeneethanol, acmc-209pku, ksc450o3l, 2,4-dichlorophenylmethyl-carbinol, 2-2,4-dichlorophenyl-ethanol
PubChem CID 2734099
IUPAC Name 2-(2,4-dichlorophenyl)ethanol
SMILES C1=CC(=C(C=C1Cl)Cl)CCO

Specifications

Color Colorless
Quantity 5 g
Formula Weight 191.05
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2,4-Dichlorophenyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.