accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 81238-58-6

IUPAC Name: 19-[2-(2,4-dinitrophenyl)hydrazin-1-ylidene]-3,6,9,12,15-pentaoxabicyclo[15.3.1]henicosa-1(20),17-dien-21-one
Synonyms: 19-[2-(2,4-dinitrophenyl)hydrazin-1-ylidene]-3,6,9,12,15-pentaoxabicyclo[15.3.1]henicosa-1(20),17-dien-21-one
Molecular Weight (g/mol): 506.47
Molecular Formula: C22H26N4O10
InChi Key: NQSRKKVRYOZHSC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=C(NN=C2C=C3COCCOCCOCCOCCOCC(=C2)C3=O)C=C1)[N+]([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results