Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

18-Crown-5 [4-(2,4-Dinitrophenylazo)phenol], TCI America™
SDP

Catalog No. C1943100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1943100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1943100MG Supplier TCI America Supplier No. C1943100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 81238-58-6
Molecular Formula C22H26N4O10
Molecular Weight (g/mol) 506.468
InChI Key NQSRKKVRYOZHSC-UHFFFAOYSA-N
Synonym 5-(2,4-Dinitrophenylazo)-2-hydroxy-1,3-xylylene-18-crown 5-Ether
PubChem CID 12866666
IUPAC Name 19-[(2,4-dinitrophenyl)hydrazinylidene]-3,6,9,12,15-pentaoxabicyclo[15.3.1]henicosa-1(20),17-dien-21-one
SMILES C1COCCOCC2=CC(=NNC3=C(C=C(C=C3)[N+](=O)[O-])[N+](=O)[O-])C=C(C2=O)COCCOCCO1

Specifications

Quantity 100 mg
Formula Weight 506.47
Physical Form Crystalline Powder
Chemical Name or Material 18-Crown-5 [4-(2,4-Dinitrophenylazo)phenol]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.