accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3NNOO

CAS RN 81282-82-8

IUPAC Name: 4-(3,5-dimethyl-1H-pyrazol-1-yl)benzoate
Synonyms: 4-(3,5-dimethylpyrazol-1-yl)benzoate
Molecular Weight (g/mol): 215.23
Molecular Formula: C12H11N2O2
InChi Key: LXNGKWBJIIXNBD-UHFFFAOYSA-M
SMILES: CC1=NN(C(C)=C1)C1=CC=C(C=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results