Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzoic acid, 97%, Thermo Scientific™

Catalog No. MO01029DA
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MO01029DA Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01029DA Supplier Thermo Scientific Chemicals Supplier No. MO01029DA

missing translation for 'validMainframeMsg'

Specifications

CAS 81282-82-8
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C12H12N2O2
Quantity 1g
MDL Number MFCD03011525
Synonym 4-3,5-dimethyl-1h-pyrazol-1-yl benzoic acid, 4-3,5-dimethylpyrazol-1-yl benzoic acid, 4-3,5-dimethyl-pyrazol-1-yl-benzoic acid, 4-3,5-dimethylpyrazolyl benzoic acid, enamine_004968, acmc-209pl6, 4-3,5-dimethyl-1-pyrazolyl benzoic acid, 4-3.5-dimethyl-pyrazol-1-yl-benzoic acid, 4-3,5-dimethyl-ih-pyrazol-i-yl benzoic acid, 4-3,5-dimethyl-1h-pyrazole-1-yl benzoic acid
InChI Key LXNGKWBJIIXNBD-UHFFFAOYSA-N
SMILES CC1=CC(=NN1C2=CC=C(C=C2)C(=O)O)C
IUPAC Name 4-(3,5-dimethylpyrazol-1-yl)benzoic acid
Molecular Weight (g/mol) 216.24
PubChem CID 693206
Formula Weight 216.24
Percent Purity 95%
Chemical Name or Material 4-(3, 5-Dimethyl-1H-pyrazol-1-yl)benzoic acid
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.