accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 813-19-4

IUPAC Name: hexabutyldistannane
Synonyms: hexabutyldistannane
Molecular Weight (g/mol): 580.12
Molecular Formula: C24H54Sn2
InChi Key: REDSKZBUUUQMSK-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)[Sn](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (6)
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)