Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexa-n-butylditin, 97%

Catalog No. AAA1200730
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1200730 250 g
AAA1200709 10 g
AAA1200718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1200730 Supplier Thermo Scientific Chemicals Supplier No. A1200730
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 813-19-4 | C24H54Sn2 | 580.12 g/mol

Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 813-19-4
Molecular Formula C24H54Sn2
Molecular Weight (g/mol) 580.12
MDL Number MFCD00009417
InChI Key REDSKZBUUUQMSK-UHFFFAOYSA-N
Synonym hexabutylditin, hexabutyldistannane, bis, distannane, hexabutyl, tin, hexabutyldi, stannane, hexabutyldi, hexa-n-butylditin, 1,1,1,2,2,2-hexabutyldistannane, hexabutyidistannane, hexabutyl distannane
PubChem CID 6327815
IUPAC Name tributyltin
SMILES CCCC[Sn](CCCC)(CCCC)[Sn](CCCC)(CCCC)CCCC

Specifications

Density 1.152
Boiling Point 197°C to 198°C (10 mmHg)
Flash Point 124°C (255°F)
Refractive Index 1.512
Linear Formula (CH3CH2CH2CH2)3Sn-Sn(CH2CH2CH2CH3)3
Quantity 250 g
UN Number UN2788
Beilstein 3605476
Sensitivity Air and moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 580.07
Percent Purity 98%
Chemical Name or Material Hexa-n-butylditin
Show More Show Less
Hazard Category H301-H311-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
EINECSNumber 212-383-3
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.