accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOHOOH3C

CAS RN 813-78-5

IUPAC Name: dimethoxyphosphinic acid
Synonyms: dimethyl phosphate
Molecular Weight (g/mol): 126.05
Molecular Formula: C2H7O4P
InChi Key: KKUKTXOBAWVSHC-UHFFFAOYSA-N
SMILES: COP(O)(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results