Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl phosphate, 98%

Catalog No. AC325130010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC325130010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC325130010 Supplier Thermo Scientific Chemicals Supplier No. 325130010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 813-78-5 | C2H7O4P | 126.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 813-78-5
Molecular Formula C2H7O4P
Molecular Weight (g/mol) 126.02
MDL Number MFCD00014887
InChI Key KKUKTXOBAWVSHC-UHFFFAOYSA-N
Synonym dimethyl phosphate, dimethylphosphate, phosphoric acid, dimethyl ester, o,o-dimethyl phosphate, unii-hi4k2c9uei, methyl phosphate meo 2 ho po, o,o-dimethyl hydrogen phosphate, hi4k2c9uei, dimethoxyphosphinic acid, dimethyl acid phosphate
PubChem CID 13134
IUPAC Name dimethyl hydrogen phosphate
SMILES COP(=O)(O)OC

Specifications

Color Brown or Colorless to Yellow
Density 1.3230g/mL
Boiling Point 174°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4080 to 1.41
Quantity 1 g
Beilstein 01,III,1205
Specific Gravity 1.323
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in ethanol,acetone
Formula Weight 126.02
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Dimethyl phosphate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-389-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.