accessibility menu, dialog, popup

UserName

Viewing profile as user:
FN

CAS RN 81329-31-9

IUPAC Name: 1-(4-fluorophenyl)-1H-pyrrole
Synonyms: 1-(4-fluorophenyl)pyrrole
Molecular Weight (g/mol): 161.18
Molecular Formula: C10H8FN
InChi Key: YMAPGGZDHAHLGN-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)N1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results