Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Fluorophenyl)pyrrole, 98%, Thermo Scientific™

Catalog No. AAA1395706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1395706 5 g
AAA1395703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1395706 Supplier Thermo Scientific Chemicals Supplier No. A1395706

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81329-31-9
Molecular Formula C10H8FN
Molecular Weight (g/mol) 161.18
MDL Number MFCD00047066
InChI Key YMAPGGZDHAHLGN-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl pyrrole, 1-4-fluorophenyl-1h-pyrrole, 1-4-fluoro-phenyl-1h-pyrrole, 4-fluorophenyl pyrrole, acmc-20apan, maybridge1_002955, n-4-fluorophenyl pyrrole, 1h-pyrrole,1-4-fluorophenyl
PubChem CID 727844
IUPAC Name 1-(4-fluorophenyl)pyrrole
SMILES FC1=CC=C(C=C1)N1C=CC=C1

Specifications

Melting Point 50°C to 52°C
Quantity 5 g
Formula Weight 161.18
Percent Purity 98%
Chemical Name or Material 1-(4-Fluorophenyl)pyrrole
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.