accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SnH3CCH3SiH3CCH3CH3

CAS RN 81353-38-0

IUPAC Name: trimethyl[2-(tributylstannyl)ethynyl]silane
Synonyms: trimethyl[2-(tributylstannyl)ethynyl]silane
Molecular Weight (g/mol): 387.27
Molecular Formula: C17H36SiSn
InChi Key: JGOIIPRSFZFFHG-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C#C[Si](C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results