Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyl(trimethylsilylethynyl)tin 96.0+%, TCI America™
SDP

Catalog No. T18655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18655G 5 g
T186525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18655G Supplier TCI America Supplier No. T18655G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 81353-38-0
Molecular Formula C17H36SiSn
Molecular Weight (g/mol) 387.27
MDL Number MFCD03844806
InChI Key JGOIIPRSFZFFHG-UHFFFAOYSA-N
Synonym 1-Tributylstannyl-2-trimethylsilylacetylene, Trimethyl(tributylstannylethynyl)silane
PubChem CID 639663
IUPAC Name trimethyl(2-tributylstannylethynyl)silane
SMILES CCCC[Sn](CCCC)(CCCC)C#C[Si](C)(C)C

Specifications

Color Yellow
Quantity 5 g
UN Number 2788
Formula Weight 387.27
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Tributyl(trimethylsilylethynyl)tin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.