accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3Br

CAS RN 81367-59-1

IUPAC Name: 1-bromo-2-methylhexadecane
Synonyms: 1-bromo-2-methylhexadecane
Molecular Weight (g/mol): 319.37
Molecular Formula: C17H35Br
InChi Key: VHNPILDWLZOLCU-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCC(C)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results