Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-methylhexadecane 98.0+%, TCI America™
SDP

Catalog No. B14561G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B14561G Supplier TCI America Supplier No. B14561G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 81367-59-1
Molecular Formula C17H35Br
Molecular Weight (g/mol) 319.371
MDL Number MFCD00142588
InChI Key VHNPILDWLZOLCU-UHFFFAOYSA-N
Synonym 2-Methylhexadecyl Bromide
PubChem CID 10757979
IUPAC Name 1-bromo-2-methylhexadecane
SMILES CCCCCCCCCCCCCCC(C)CBr

Specifications

Color Yellow
Quantity 1 g
Formula Weight 319.37
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-methylhexadecane
EINECSNumber (2)-0067
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.