accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CPOH3CH3C

CAS RN 814-29-9

IUPAC Name: 1-(dibutylphosphoryl)butane
Synonyms: tributylphosphine oxide
Molecular Weight (g/mol): 218.32
Molecular Formula: C12H27OP
InChi Key: MNZAKDODWSQONA-UHFFFAOYSA-N
SMILES: CCCCP(=O)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)