accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 814-87-9

IUPAC Name: dialuminium(3+) trioxalate
Synonyms: dialuminium(3+) trioxalate
Molecular Weight (g/mol): 318.02
Molecular Formula: C6Al2O12
InChi Key: ZCLVNIZJEKLGFA-UHFFFAOYSA-H
SMILES: [Al+3].[Al+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)