Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Oxalate, MP Biomedicals

Catalog No. ICN206473 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN206473 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN206473 Supplier MP Biomedicals, Inc Supplier No. 0520647310
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 814-87-9
Molecular Formula C6Al2O12
Molecular Weight (g/mol) 318.02
MDL Number MFCD00050791
InChI Key ZCLVNIZJEKLGFA-UHFFFAOYSA-H
Synonym aluminum oxalate, dialuminium trioxalate, unii-dq79q766vn, aluminum, tris ethanedioato 2-di, dialuminum oxalate, aluminum oxalate hydrate, dialuminium 3+ trioxalate ion, dialuminium 3+ trioxalate, aluminum oxalate hydrate 10g, dialuminium 3+ ion trioxalate
PubChem CID 93561
IUPAC Name dialuminium(3+) trioxalate
SMILES [Al+3].[Al+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

Quantity 10 g
Formula Weight 318.03
Chemical Name or Material Aluminum Oxalate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.